Läs mer
Methyl 2-(3-Nitrobenzylidene)-3-oxobutanoate (1:1 E/Z Mixture), TRC
Produktkod. 30475579
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30475579 5 g
1 options
Produktkod. 30475579 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3031255G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 2-(3-Nitrobenzylidene)-3-oxobutanoate (1:1 E/Z Mixture)
CAS 39562-17-9
Molekylformel C12H11NO5
Kvantitet 5 g
Synonym Methyl 2-(3-Nitrobenzylidene)acetoacetate, Methyl 2-(m-Nitrobenzylidene)acetoacetate, Methyl 3-Nitrobenzylideneacetoacetate, Methyl alpha-Acetyl-beta-(m-nitrophenyl)acrylate
InChI formel InChI=1S/C12H11NO5/c1-8(14)11(12(15)18-2)7-9-4-3-5-10(6-9)13(16)17/h3-7H,1-2H3/b11-7-
LEDER COC(=O)\C(=C/c1cccc(c1)[N+](=O)[O-])\C(=O)C
IUPAC-namn methyl (2Z)-2-[(3-nitrophenyl)methylidene]-3-oxobutanoate
Molekylvikt (g/mol) 249.22
Formel vikt 249.0637