Läs mer
Methyl 2,3-O-Isopropylidene-5-O-p-tolylsulfonyl-ß-D-ribofuranoside, TRC
Produktkod. 30473068
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30473068 5 g
1 options
Produktkod. 30473068 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3147005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 2,3-O-Isopropylidene-5-O-p-tolylsulfonyl-beta-D-ribofuranoside
CAS 4137-56-8
Molekylformel C16 H22 O7 S
Kvantitet 5 g
Synonym Methyl-2,3-O-isopropylidene-5-O-tosyl-beta-D-ribofuranoside','Furo[3,4-d]-1,3-dioxole','β-D-ribofuranoside deriv. (ZCI), Ribofuranoside','methyl 2,3-O-isopropylidene-','p-toluenesulfonate (6CI','7CI), Ribofuranoside','methyl 2,3-O-isopropylidene-','p-toluenesulfonate','β-D- (8CI), β-D-Ribofuranoside','methyl 2,3-O-(1-methylethylidene)-','4-methylbenzenesulfonate (9CI), Methyl 2,3-O-isopropylidene-5-O-p-toluenesulfonyl-β-D-ribofuranoside, Methyl 2,3-O-isopropylidene-5-O-p-tolylsulfonyl-β-D-ribofuranoside, Methyl 2,3-O-isopropylidene-5-O-tosyl-D-ribonucleoside, Methyl 2,3-O-isopropylidene-5-O-tosyl-β-D-ribofuranoside, Methyl 2,3-O-isopropylidene-5-O-β-toluenesulfonyl-β-D-ribofuranoside, NSC 85192
InChI formel InChI=1S/C16H22O7S/c1-10-5-7-11(8-6-10)24(17,18)20-9-12-13-14(15(19-4)21-12)23-16(2,3)22-13/h5-8,12-15H,9H2,1-4H3/t12-,13-,14-,15-/m1/s1
LEDER CO[C@@H]1O[C@H](COS(=O)(=O)c2ccc(C)cc2)[C@H]3OC(C)(C)O[C@@H]13
IUPAC-namn [(3aR,4R,6R,6aR)-4-methoxy-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]methyl 4-methylbenzenesulfonate
Molekylvikt (g/mol) 358.41
Formel vikt 358.1086