Läs mer
Methyl 2-[4-[(1RS)-1-Hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]-butyl]phenyl]-2-methylpropanoate (Fexofenadine Methyl Ester), Mikromol™
Produktkod. 30052190
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052190 100 mg
30051886 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052190 Leverantör Mikromol Leverantörsnummer MM0507.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 2-[4-[(1RS)-1-Hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]-butyl]phenyl]-2-methylpropanoate (Fexofenadine Methyl Ester)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 2-[4-[(1RS)-1-Hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]-butyl]phenyl]-2-methylpropanoate
CAS 154825-96-4
Analyt- eller komponentnamn Methyl 2-[4-[(1RS)-1-Hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]-butyl]phenyl]-2-methylpropanoate
Frakt skick Room Temperature
Molekylformel C33 H41 N O4
Certifikat och efterlevnad ISO 17025
Synonym Fexofenadine Hydrochloride Imp. D (EP), Fexofenadine Imp. D (EP), Methyl 2-[4-[(1RS)-1-Hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]-butyl]phenyl]-2-methylpropanoate, Fexofenadine Hydrochloride Impurity D, Fexofenadine Impurity D
LEDER COC(=O)C(C)(C)c1ccc(cc1)C(O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4
Molekylvikt (g/mol) 515.69
Kvantitet 100 mg
Formel vikt 515.694 g/mol