Läs mer
Methyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate, Mikromol™
Produktkod. 30462746
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30462746 4 x 25 mg
17903186 100 mg
17950706 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30462746 Leverantör Mikromol Leverantörsnummer MM0063.0400254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
CAS 2157488-46-3
Analyt- eller komponentnamn Methyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
Frakt skick Room Temperature
Molekylformel C20 H22 Cl N O4
Certifikat och efterlevnad ISO 17025
Synonym Bezafibrate Imp. C (EP), Methyl 2-[4-[2-[(4-Chloro-benzoyl)amino]ethyl]phenoxy]-2-methylpropanoate, Bezafibrate Impurity C
InChI formel InChI=1S/C17H14N4O4/c1-25-17(24)21-16-19-13-7-4-11(8-14(13)20-16)15(23)10-2-5-12(6-3-10)18-9-22/h2-9H,1H3,(H,18,22)(H2,19,20,21,24)
Lösningstyp Neat
LEDER COC(=O)C(C)(C)Oc1ccc(CCNC(=O)c2ccc(Cl)cc2)cc1
IUPAC-namn methyl 2-[4-[2-[(4-chlorobenzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
Molekylvikt (g/mol) 375.85
Kvantitet 4 x 25 mg
Formel vikt 375.1237 g/mol