missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2,6-dichloropyridine-4-carboxylate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediates.
Solubility
Sparingly Soluble (0.13 g/L) (25°C)
Notes
Keep away from oxidizing agents, reducing agents, bases, amines. Store in cool place.
It is used as pharmaceutical intermediates.
Solubility
Sparingly Soluble (0.13 g/L) (25°C)
Notes
Keep away from oxidizing agents, reducing agents, bases, amines. Store in cool place.
Specifications
Melting Point | 80°C to 84°C |
Quantity | 1 g |
Synonym | methyl 2,6-dichloroisonicotinate, methyl2,6-dichloroisonicotinate, 2,6-chloro-4-pyridinecarboxylic acid methyl ester, 2,6-dichloroisonicotinic acid methyl ester, methyl-2,6-dichloroisonicotinate, 4-pyridinecarboxylic acid, 2,6-dichloro-, methyl ester, methyl-2,6-dichlorisonicotinat, acmc-209yt4, xskghsuhoyebtk-uhfffaoysa, methyl 2,6-dichloro-isonicotinate |
Solubility Information | Sparingly Soluble (0.13g/L) (25°C) |
Formula Weight | 206.03 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 2,6-dichloropyridine-4-carboxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'