Läs mer
Methyl 2-Acetamido-2-deoxy-a-D-glucopyranoside, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl 2-Acetamido-2-deoxy-a-D-glucopyranoside |
| CAS | 6082-04-8 |
| Molekylformel | C9 H17 N O6 |
| Kvantitet | 25 mg |
| Synonym | α-D-Glucopyranoside','methyl 2-(acetylamino)-2-deoxy-, Glucopyranoside','methyl 2-acetamido-2-deoxy-','α-D- (8CI), Methyl 2-(acetylamino)-2-deoxy-α-D-glucopyranoside, Methyl 2-N-acetamido-2-deoxy-α-D-glucopyranoside, Methyl 2-acetamido-2-deoxy-α-D-glucopyranoside, Methyl 2-acetamido-2-deoxy-α-D-glucoside, Methyl 2-deoxy-2-acetamido-α-D-glucopyranoside, Methyl N-acetyl-α-D-glucosaminide, Methyl N-acetyl-α-glucosaminide, NSC 77914, α-Methyl N-acetyl-D-glucosaminide, α-Methyl N-acetylglucosaminide |
| InChI formel | InChI=1S/C9H17NO6/c1-4(12)10-6-8(14)7(13)5(3-11)16-9(6)15-2/h5-9,11,13-14H,3H2,1-2H3,(H,10,12)/t5-,6-,7-,8-,9+/m1/s1 |
| LEDER | CO[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(=O)C |
| IUPAC-namn | N-[(2S,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-methoxyoxan-3-yl]acetamide |
| Molekylvikt (g/mol) | 235.23 |
| Formel vikt | 235.1056 |