Läs mer
Methyl 2-Acetamido-4-O-(2-acetamido-2-deoxy-Beta-D-gluco- pyranosyl)-2-deoxy-6-O-(Alpha-L-fucopyranosyl)-Beta-D-glucopyranoside, TRC
Produktkod. 30464466
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464466 25 mg
30471453 50 mg
30479474 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30464466 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM27250025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 97242-84-7
Molekylformel C23 H40 N2 O15
Molekylvikt (g/mol) 584.57
IUPAC-namn N-[(2S,3R,4R,5S,6R)-2-[(2R,3S,4R,5R,6R)-5-acetamido-4-hydroxy-6-methoxy-2-[[(2R,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-3-yl]oxy-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
LEDER CO[C@@H]1O[C@H](CO[C@@H]2O[C@@H](C)[C@@H](O)[C@@H](O)[C@@H]2O)[C@@H](O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3NC(=O)C)[C@H](O)[C@H]1NC(=O)C
Specifikationer
Kemiskt namn eller material Methyl 2-Acetamido-4-O-(2-acetamido-2-deoxy-beta-D-gluco-pyranosyl)-2-deoxy-6-O-(alpha-L-fucopyranosyl)-beta-D-glucopyranoside
Kvantitet 25 mg
InChI formel InChI=1S/C23H40N2O15/c1-7-14(29)18(33)19(34)23(37-7)36-6-11-20(17(32)13(25-9(3)28)21(35-4)39-11)40-22-12(24-8(2)27)16(31)15(30)10(5-26)38-22/h7,10-23,26,29-34H,5-6H2,1-4H3,(H,24,27)(H,25,28)/t7-,10+,11+,12+,13+,14+,15+,16+,17+,18+,19-,20+,21+,22-,23+/m0/s1
Formel vikt 584.2429