Läs mer
Methyl 2-Amino-4-fluoro-3-methylbenzoate, TRC
Produktkod. 30479179
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479179 10 mg
30474854 50 mg
30478812 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479179 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM28865810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 928839-58-1
Molekylformel C9H10NO2F
Molekylvikt (g/mol) 183.17
IUPAC-namn methyl 2-amino-4-fluoro-3-methylbenzoate
LEDER COC(=O)c1ccc(F)c(C)c1N
Specifikationer
Kemiskt namn eller material Methyl 2-Amino-4-fluoro-3-methylbenzoate
Kvantitet 10 mg
InChI formel InChI=1S/C9H10FNO2/c1-5-7(10)4-3-6(8(5)11)9(12)13-2/h3-4H,11H2,1-2H3
Formel vikt 183.0696