Läs mer
Methyl 2-Amino-5-methylbenzoate, TRC
Produktkod. 30480490
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30480490 25 g
1 options
Produktkod. 30480490 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM28688525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 2-Amino-5-methylbenzoate
CAS 18595-16-9
Molekylformel C9 H11 N O2
Kvantitet 25 g
Synonym Benzoic acid','2-amino-5-methyl-','methyl ester, m-Toluic acid','6-amino-','methyl ester (6CI,8CI), 2-Amino-5-methylbenzoic acid methyl ester, Methyl 2-amino-5-methylbenzoate, Methyl 5-methylanthranilate
InChI formel InChI=1S/C9H11NO2/c1-6-3-4-8(10)7(5-6)9(11)12-2/h3-5H,10H2,1-2H3
LEDER COC(=O)c1cc(C)ccc1N
IUPAC-namn methyl 2-amino-5-methylbenzoate
Molekylvikt (g/mol) 165.19
Formel vikt 165.079