missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-aminopyridine-4-carboxylate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Methyl 2-aminopyridine-4-carboxylate is an inhibitor of nitrogen monoxide synthetase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 2-aminopyridine-4-carboxylate is an inhibitor of nitrogen monoxide synthetase.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Methyl 2-aminopyridine-4-carboxylate is an inhibitor of nitrogen monoxide synthetase.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 145°C to 149°C |
Quantity | 1 g |
Beilstein | 128642 |
Solubility Information | Insoluble in water. |
Formula Weight | 152.15 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 2-aminopyridine-4-carboxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'