Läs mer
Methyl 2-Benzamidoacetate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl 2-Benzamidoacetate |
| CAS | 1205-08-9 |
| Molekylformel | C10 H11 N O3 |
| Kvantitet | 100 mg |
| Synonym | Methyl N-Benzoylglycinate, Methyl Benzoylaminoacetate, Methyl Benzoylglycinate, Methyl Hippurate, N-Benzoylglycine Methyl Ester |
| InChI formel | InChI=1S/C10H11NO3/c1-14-9(12)7-11-10(13)8-5-3-2-4-6-8/h2-6H,7H2,1H3,(H,11,13) |
| LEDER | COC(=O)CNC(=O)c1ccccc1 |
| IUPAC-namn | methyl 2-benzamidoacetate |
| Molekylvikt (g/mol) | 193.199 |
| Formel vikt | 193.074 |