Learn More
Methyl 2-bromo-3-methylbenzoate, 98%, Thermo Scientific Chemicals
Catalog Number p-7054000
Click to view available options
Quantity:
25 g
100 g
131001-86-0
C9H9BrO2
229.07
MFCD00144762
QAOFGUXVDAZKBW-UHFFFAOYSA-N
2-bromo-3-methyl-benzoic acid methyl ester, methyl2-bromo-3-methylbenzoate, benzoic acid, 2-bromo-3-methyl-, methyl ester, 2-bromo-3-methylbenzoic acid methyl ester, methyl 2-bromo-3-methyl-benzoate, pubchem3966, acmc-1c3gk, methyl 2-bromo-m-toluate, intermediates-zcf02010, ksc495c9h
10823279
methyl 2-bromo-3-methylbenzoate
COC(=O)C1=C(Br)C(C)=CC=C1
131001-86-0
C9H9BrO2
229.07
MFCD00144762
QAOFGUXVDAZKBW-UHFFFAOYSA-N
2-bromo-3-methyl-benzoic acid methyl ester, methyl2-bromo-3-methylbenzoate, benzoic acid, 2-bromo-3-methyl-, methyl ester, 2-bromo-3-methylbenzoic acid methyl ester, methyl 2-bromo-3-methyl-benzoate, pubchem3966, acmc-1c3gk, methyl 2-bromo-m-toluate, intermediates-zcf02010, ksc495c9h
10823279
methyl 2-bromo-3-methylbenzoate
COC(=O)C1=C(Br)C(C)=CC=C1
Methyl 2-bromo-3-methylbenzoate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 2-bromo-3-methylbenzoate is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Color Red
Quantity 25 g
Beilstein 4177165
Solubility Information Slightly soluble in water.
Formula Weight 229.07
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 2-bromo-3-methylbenzoate