Läs mer
Methyl-2-bromo-4-bromomethylbenzoate, TRC
Produktkod. 30465568
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30465568 500 mg
30473179 100 mg
30471488 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30465568 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM296273500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 128577-48-0
Molekylformel C9H8Br2O2
Molekylvikt (g/mol) 307.97
IUPAC-namn methyl 2-bromo-4-(bromomethyl)benzoate
LEDER COC(=O)c1ccc(CBr)cc1Br
Specifications
Kemiskt namn eller material Methyl-2-bromo-4-bromomethylbenzoate
Kvantitet 500 mg
InChI formel InChI=1S/C9H8Br2O2/c1-13-9(12)7-3-2-6(5-10)4-8(7)11/h2-4H,5H2,1H3
Formel vikt 305.8891