Läs mer
Methyl 2-Bromo-5-chloronicotinate, TRC
Produktkod. 30471486
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30471486 250 mg
30483767 100 mg
30465801 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30471486 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM296720250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1214324-95-4
Molekylformel C7H5NO2ClBr
Molekylvikt (g/mol) 250.47
Synonym Methyl 2-Bromo-5-chloro-3-pyridinecarboxylate
LEDER COC(=O)c1cc(Cl)cnc1Br
Specifikationer
Kemiskt namn eller material Methyl 2-Bromo-5-chloronicotinate
Kvantitet 250 mg
InChI formel InChI=1S/C7H5BrClNO2/c1-12-7(11)5-2-4(9)3-10-6(5)8/h2-3H,1H3
Formel vikt 248.9192