Learn More
Methyl 2-(bromomethyl)acrylate, 96%, Thermo Scientific Chemicals
Catalog Number p-4510682
Click to view available options
Quantity:
1 g
5 g
25 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4224-69-5
C5H7BrO2
179.01
MFCD00011697
CFTUQSLVERGMHL-UHFFFAOYSA-N
methyl 2-bromomethyl acrylate, methyl 2-bromomethyl prop-2-enoate, methyl 2-bromomethyl-2-propenoate, methyl alpha-bromomethyl acrylate, 2-bromomethyl methyl acrylate, 2-propenoic acid, 2-bromomethyl-, methyl ester, 2-bromomethyl acrylic acid methyl ester, acrylic acid, 2-bromomethyl-, methyl ester, zlchem 195, pubchem17371
521093
methyl 2-(bromomethyl)prop-2-enoate
COC(=O)C(=C)CBr
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4224-69-5
C5H7BrO2
179.01
MFCD00011697
CFTUQSLVERGMHL-UHFFFAOYSA-N
methyl 2-bromomethyl acrylate, methyl 2-bromomethyl prop-2-enoate, methyl 2-bromomethyl-2-propenoate, methyl alpha-bromomethyl acrylate, 2-bromomethyl methyl acrylate, 2-propenoic acid, 2-bromomethyl-, methyl ester, 2-bromomethyl acrylic acid methyl ester, acrylic acid, 2-bromomethyl-, methyl ester, zlchem 195, pubchem17371
521093
methyl 2-(bromomethyl)prop-2-enoate
COC(=O)C(=C)CBr
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Brown to Colorless
Density 1.4890g/mL
Boiling Point 35.0°C to 37.0°C (13.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4870 to 1.4890
Quantity 1 g
Specific Gravity 1.489
Formula Weight 179.01
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Methyl 2-(bromomethyl)acrylate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 2-(bromomethyl)acrylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.