Läs mer
Methyl 2-Bromomethylbenzoate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl 2-Bromomethylbenzoate |
| CAS | 2417-73-4 |
| Molekylformel | C9 H9 Br O2 |
| Kvantitet | 5 g |
| Synonym | 2-Bromomethylbenzoic Acid Methyl Ester, 2-Bromomethylbenzoic Methyl Ester, 2-Carbomethoxy-α-bromotoluene, 2-Carbomethoxybenzyl Bromide, 2-Methoxycarbonylbenzyl Bromide, Methyl 2-(Bromomethyl)benzoate, Methyl o-(Bromomethyl)benzoate, Methyl α-Bromo-o-toluate, o-Carbomethoxybenzyl Bromide, α-Bromo-o-Toluic Acid Methyl Ester |
| InChI formel | InChI=1S/C9H9BrO2/c1-12-9(11)8-5-3-2-4-7(8)6-10/h2-5H,6H2,1H3 |
| LEDER | COC(=O)c1ccccc1CBr |
| IUPAC-namn | methyl 2-(bromomethyl)benzoate |
| Molekylvikt (g/mol) | 229.07 |
| Formel vikt | 227.9786 |