Läs mer
Methyl-2-bromopentanoate, TRC
Produktkod. 30468444
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30468444 10 g
30472829 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30468444 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26510010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19129-92-1
Molekylformel C6H11BrO2
Molekylvikt (g/mol) 195.05
Synonym (±)-Methyl 2-Bromopentanoate, 2-Bromovaleric Acid Methyl Ester, Methyl 2-Bromopentanoate, Methyl alpha-Bromopentanoate, Methyl alpha-Bromovalerate
IUPAC-namn methyl 2-bromopentanoate
LEDER CCCC(Br)C(=O)OC
Specifikationer
Kemiskt namn eller material Methyl-2-bromopentanoate
Kvantitet 10 g
InChI formel InChI=1S/C6H11BrO2/c1-3-4-5(7)6(8)9-2/h5H,3-4H2,1-2H3
Formel vikt 193.9942