missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(chloromethyl)-1,3-oxazole-4-carboxylate, 97%, Thermo Scientific™

Catalog Number p-3111006
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
208465-72-9
C6H6ClNO3
175.57
MFCD06660133
CMUKPCIZFMTLKD-UHFFFAOYSA-N
methyl 2-chloromethyl oxazole-4-carboxylate, methyl 2-chloromethyl-1,3-oxazole-4-carboxylate, 4-oxazolecarboxylic acid, 2-chloromethyl-, methyl ester, methyl 2-chloromethyl-4-oxazolecarboxylate, pubchem22375, 2-chloromethyl-5-methyloxazole-4-carboxylate, 2-chloromethyloxazole-4-carboxylic acid methyl ester, methyl 2-chloro-methyl-1,3-oxazole-4-carboxylate
2763199
methyl 2-(chloromethyl)-1,3-oxazole-4-carboxylate
COC(=O)C1=COC(CCl)=N1
This item is not returnable. View return policy
208465-72-9
C6H6ClNO3
175.57
MFCD06660133
CMUKPCIZFMTLKD-UHFFFAOYSA-N
methyl 2-chloromethyl oxazole-4-carboxylate, methyl 2-chloromethyl-1,3-oxazole-4-carboxylate, 4-oxazolecarboxylic acid, 2-chloromethyl-, methyl ester, methyl 2-chloromethyl-4-oxazolecarboxylate, pubchem22375, 2-chloromethyl-5-methyloxazole-4-carboxylate, 2-chloromethyloxazole-4-carboxylic acid methyl ester, methyl 2-chloro-methyl-1,3-oxazole-4-carboxylate
2763199
methyl 2-(chloromethyl)-1,3-oxazole-4-carboxylate
COC(=O)C1=COC(CCl)=N1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 175.57
Percent Purity 97%
Chemical Name or Material Methyl 2-(chloromethyl)-1, 3-oxazole-4-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'