Läs mer
Methyl 2-Cyano-2-oximinoacetate, TRC
Produktkod. 30473539
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30473539 100 mg
1 options
Produktkod. 30473539 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM294603100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 2-Cyano-2-oximinoacetate
CAS 61295-92-9
Molekylformel C4H4N2O3
Kvantitet 100 mg
Synonym 2-Cyano-2-(hydroxyimino)-acetic Acid Methyl Ester, Cyano(hydroxyimino)-acetic Acid Methyl Ester, NSC 158573
InChI formel InChI=1S/C4H4N2O3/c1-9-4(7)3(2-5)6-8/h8H,1H3/b6-3+
LEDER COC(=O)\C(=N\O)\C#N
IUPAC-namn methyl (2E)-2-cyano-2-hydroxyiminoacetate
Molekylvikt (g/mol) 128.09
Formel vikt 128.0222