Läs mer
Methyl 2-Cyanomethylbenzoate, Mikromol™
Produktkod. 30051438
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051438 100 mg
30050900 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051438 Leverantör Mikromol Leverantörsnummer MM0001.31
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 2-Cyanomethylbenzoate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 2-Cyanomethylbenzoate
CAS 4-6-5597
Analyt- eller komponentnamn Methyl 2-Cyanomethylbenzoate
Frakt skick Room Temperature
Molekylformel C10 H9 N O2
Certifikat och efterlevnad ISO 17025
Synonym 2-(Cyanomethyl)-Benzoic acid methyl ester, Methyl 2-(cyanomethyl)benzoate, o-Toluic acid, alpha-cyano-, methyl ester (6CI,7CI,8CI)
InChI formel InChI=1S/C10H9NO2/c1-13-10(12)9-5-3-2-4-8(9)6-7-11/h2-5H,6H2,1H3
LEDER COC(=O)c1ccccc1CC#N
IUPAC-namn methyl 2-(cyanomethyl)benzoate
Molekylvikt (g/mol) 175.18
Kvantitet 100 mg
Formel vikt 175.0633 g/mol