Läs mer
Methyl 2-Cyclohexyl-2-hydroxyphenylacetate, TRC
Produktkod. 30476346
Change view
Kvantitet:
500 mg
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476346 500 mg
30478184 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30476346 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM294645500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 10399-13-0
Molekylformel C15 H20 O3
Molekylvikt (g/mol) 248.32
Synonym Methyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate, Phenylcyclohexylglycolic Acid Methyl Ester
IUPAC-namn methyl 2-cyclohexyl-2-hydroxy-2-phenylacetate
LEDER COC(=O)C(O)(C1CCCCC1)c2ccccc2
Specifikationer
Kemiskt namn eller material Phenylcyclohexylglycolic Acid Methyl Ester
Kvantitet 500 mg
InChI formel InChI=1S/C15H20O3/c1-18-14(16)15(17,12-8-4-2-5-9-12)13-10-6-3-7-11-13/h2,4-5,8-9,13,17H,3,6-7,10-11H2,1H3
Formel vikt 248.1412