Läs mer
Methyl 2-Ethoxy-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-benzimidazole-7-carboxylate (Candesartan Methyl Ester), Mikromol™
Produktkod. 30051155
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051155 100 mg
30050781 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051155 Leverantör Mikromol Leverantörsnummer MM0776.10
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 2-Ethoxy-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-benzimidazole-7-carboxylate (Candesartan Methyl Ester)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Candesartan Methyl Ester
CAS 139481-69-9
Analyt- eller komponentnamn Candesartan Methyl Ester
Frakt skick Room Temperature
Molekylformel C25 H22 N6 O3
Certifikat och efterlevnad ISO 17025
Synonym Candesartan Cilexetil Imp. I (EP), 1H-Benzimidazole-7-carboxylic acid, 2-ethoxy-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, methyl ester, 1H-Benzimidazole-7-carboxylic acid, 2-ethoxy-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, methyl ester (9CI), Methyl 1-[(2'-(1H-tetrazol-5-yl)biphenyl-4-yl)methyl]-2-ethoxybenzimidazole-7-carboxylate, Methyl 2-ethoxy-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]benzimidazole-7-carboxylate, Candesartan Cilexetil Impurity I
LEDER CCOc1nc2cccc(C(=O)OC)c2n1Cc3ccc(cc3)c4ccccc4c5nnn[nH]5
Molekylvikt (g/mol) 454.49
Kvantitet 100 mg
Formel vikt 454.49 g/mol