Läs mer
Methyl 2-fluoro-3-nitrobenzoate, TRC
Produktkod. 30477088
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30477088 250 mg
30468602 500 mg
30465814 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30477088 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM305423250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 946126-94-9
Molekylformel C8H6FNO4
Molekylvikt (g/mol) 199.14
IUPAC-namn methyl 2-fluoro-3-nitrobenzoate
LEDER COC(=O)c1cccc(c1F)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material Methyl 2-fluoro-3-nitrobenzoate
Kvantitet 250 mg
InChI formel InChI=1S/C8H6FNO4/c1-14-8(11)5-3-2-4-6(7(5)9)10(12)13/h2-4H,1H3
Formel vikt 199.0281