Läs mer
Methyl 2-Hydroxy-2-methoxyacetate, Technical Grade, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl 2-Hydroxy-2-methoxyacetate |
| CAS | 19757-97-2 |
| Molekylformel | C4H8O4 |
| Kvantitet | 500 mg |
| Synonym | Methoxyglycolic Acid Methyl Ester, Glyoxylic Acid Methyl Ester Methyl Hemiacetal, 2-Hydroxy-2-methoxyacetic Acid Methyl Ester, GMHA, Glycolic Acid Methyl Ester Hemiacetal, Methyl 2-Hydroxy-2-methoxyacetate, Methyl Glyoxylate Methyl Hemiacetal, α-Hydroxy-α-Methoxyacetic Acid |
| InChI formel | InChI=1S/C4H8O4/c1-7-3(5)4(6)8-2/h3,5H,1-2H3 |
| LEDER | COC(O)C(=O)OC |
| Molekylvikt (g/mol) | 120.1 |
| Formel vikt | 120.0423 |