Läs mer
Methyl 2-Hydroxy-5-methylbenzoate, TRC
Produktkod. 30330207
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30330207 500 mg
1 options
Produktkod. 30330207 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB585318500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 2-Hydroxy-5-methylbenzoate
CAS 22717-57-3
Molekylformel C9H10O3
Kvantitet 500 mg
Synonym 2-(Methoxycarbonyl)-4-methylphenol, 2-Hydroxy-5-methylbenzoic Acid Methyl Ester, Methyl 5-Methylsalicylate, NSC 165635
InChI formel InChI=1S/C9H10O3/c1-6-3-4-8(10)7(5-6)9(11)12-2/h3-5,10H,1-2H3
LEDER COC(=O)c1cc(C)ccc1O
IUPAC-namn methyl 2-hydroxy-5-methylbenzoate
Molekylvikt (g/mol) 166.18
Formel vikt 166.063