missing translation for 'onlineSavingsMsg'
Learn More

Methyl-2-isocyanatobenzoate, 97%, Thermo Scientific Chemicals

Catalog Number p-4259018
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1793-07-3
C9H7NO3
177.16
ATFKBWNEVZUSKC-UHFFFAOYSA-N
2-carbomethoxyphenyl isocyanate, 2-methoxycarbonylphenyl isocyanate, 2-methoxycarbonyl phenyl isocyanate, 2-carbomethoxyphenylisocyanate, 2-methoxycarbonyl phenylisocyanate, benzoic acid, 2-isocyanato-, methyl ester, acmc-1bqmd, methyl 2 isocyanatobenzoate, methyl-2-isocyanatobenzoate, #
519494
methyl 2-isocyanatobenzoate
COC(=O)C1=CC=CC=C1N=C=O
This item is not returnable. View return policy
1793-07-3
C9H7NO3
177.16
ATFKBWNEVZUSKC-UHFFFAOYSA-N
2-carbomethoxyphenyl isocyanate, 2-methoxycarbonylphenyl isocyanate, 2-methoxycarbonyl phenyl isocyanate, 2-carbomethoxyphenylisocyanate, 2-methoxycarbonyl phenylisocyanate, benzoic acid, 2-isocyanato-, methyl ester, acmc-1bqmd, methyl 2 isocyanatobenzoate, methyl-2-isocyanatobenzoate, #
519494
methyl 2-isocyanatobenzoate
COC(=O)C1=CC=CC=C1N=C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 47.0°C to 50.0°C
Color White
Boiling Point 99.0°C (0.5 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 177.16
Percent Purity 97%
Physical Form Crystalline Solid or Solid
Chemical Name or Material Methyl-2-isocyanatobenzoate
compliance-icons compliance-icons
Product Identifier
  • Methyl-2-isocyanatobenzoate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'