Läs mer
Methyl 2-methoxy-5-(methylsulfonyl)benzoate, TRC
Produktkod. 30469137
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30469137 5 g
1 options
Produktkod. 30469137 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3235255G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 2-Methoxy-5-(methylsulfonyl)benzoate
CAS 63484-12-8
Molekylformel C10 H12 O5 S
Kvantitet 5 g
Synonym 2-Methoxy-5-(methylsulfonyl)benzoic acid methyl ester, Methyl 2-Methoxy-5-(methylsulfonyl)-benzoate, Tiapride Hydrochloride Imp. A (EP)
InChI formel InChI=1S/C10H12O5S/c1-14-9-5-4-7(16(3,12)13)6-8(9)10(11)15-2/h4-6H,1-3H3
LEDER COC(=O)c1cc(ccc1OC)S(=O)(=O)C
IUPAC-namn methyl 2-methoxy-5-methylsulfonylbenzoate
Molekylvikt (g/mol) 244.26
Formel vikt 244.0405