Läs mer
Methyl 2-Methylnicotinate, TRC
Produktkod. 30466411
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30466411 10 g
1 options
Produktkod. 30466411 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32302510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 2-Methylnicotinate
CAS 65719-09-7
Molekylformel C8H9NO2
Kvantitet 10 g
Synonym 2-Methyl-3-pyridinecarboxylic Acid Methyl Ester, Methyl 2-Methyl-3-pyridinecarboxylate, 2-Methylnicotinic Acid Methyl Ester
InChI formel InChI=1S/C8H9NO2/c1-6-7(8(10)11-2)4-3-5-9-6/h3-5H,1-2H3
LEDER COC(=O)c1cccnc1C
IUPAC-namn methyl 2-methylpyridine-3-carboxylate
Molekylvikt (g/mol) 151.16
Formel vikt 151.0633