missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-methylpropionimidate hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.
Solubility
Slightly soluble in water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.
Solubility
Slightly soluble in water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Color | White |
Linear Formula | (CH3)2CHC(=NH)OCH3·HCl |
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 137.61 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | Methyl 2-methylpropionimidate hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'