Learn More
Methyl 2-O-Benzoyl-4,6-di-O-benzylidene-Alpha-D-glucopyranoside, TRC
Product Code. 30469611
Change view
Click to view available options
Kvantitet:
500 mg
Product Code. Kvantitet
30469611 500 mg
1 options
Product Code. 30469611 Supplier Toronto Research Chemicals Supplier No. TRCM288965500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Methyl 2-O-Benzoyl-4,6-di-O-benzylidene-alpha-D-glucopyranoside
CAS 28642-64-0
Molekylformel C21H22O7
Kvantitet 500 mg
Synonym Methyl 4,6-O-(Phenylmethylene)-alpha-D-glucopyranoside 2-Benzoate
InChI formel InChI=1S/C21H22O7/c1-24-21-18(27-19(23)13-8-4-2-5-9-13)16(22)17-15(26-21)12-25-20(28-17)14-10-6-3-7-11-14/h2-11,15-18,20-22H,12H2,1H3/t15-,16+,17-,18-,20-,21+/m1/s1
LEDER CO[C@H]1O[C@@H]2CO[C@H](O[C@H]2[C@H](O)[C@H]1OC(=O)c3ccccc3)c4ccccc4
IUPAC-namn [(2R,4aR,6S,7R,8S,8aS)-8-hydroxy-6-methoxy-2-phenyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl] benzoate
Molekylvikt (g/mol) 386.4
Formel vikt 386.1366