missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-oxoindane-1-carboxylate, 97%, Thermo Scientific Chemicals

Catalog Number p-7038546
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
104620-34-0
C11H10O3
190.198
MFCD06200715
RWSYHHRDYRDVQL-UHFFFAOYSA-N
methyl 2-oxo-1-indanecarboxylate, methyl 2-oxoindane-1-carboxylate, methyl2-oxo-1-indanecarboxylate, methyl 2-oxo-2,3-dihydro-1h-indene-1-carboxylate, 1h-indene-1-carboxylic acid, 2,3-dihydro-2-oxo-, methyl ester, acmc-20agv9, 1-methoxycarbonyl-2-indanone, 2-oxo-indan-1-carboxylic acid methyl ester, 2-oxo-1,3-dihydro-indene-1-carboxylic acid methyl ester
10888732
methyl 2-oxo-1,3-dihydroindene-1-carboxylate
COC(=O)C1C(=O)CC2=CC=CC=C12
This item is not returnable. View return policy
104620-34-0
C11H10O3
190.198
MFCD06200715
RWSYHHRDYRDVQL-UHFFFAOYSA-N
methyl 2-oxo-1-indanecarboxylate, methyl 2-oxoindane-1-carboxylate, methyl2-oxo-1-indanecarboxylate, methyl 2-oxo-2,3-dihydro-1h-indene-1-carboxylate, 1h-indene-1-carboxylic acid, 2,3-dihydro-2-oxo-, methyl ester, acmc-20agv9, 1-methoxycarbonyl-2-indanone, 2-oxo-indan-1-carboxylic acid methyl ester, 2-oxo-1,3-dihydro-indene-1-carboxylic acid methyl ester
10888732
methyl 2-oxo-1,3-dihydroindene-1-carboxylate
COC(=O)C1C(=O)CC2=CC=CC=C12

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.
TRUSTED_SUSTAINABILITY
Melting Point 63°C to 70°C
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 190.2
Percent Purity 97%
Chemical Name or Material Methyl 2-oxoindane-1-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'