Läs mer
Methyl (2-Oxopyrrolidin-1-yl)acetate, LoGiCal
Produktkod. 30461133
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30461133 10 mg
1 options
Produktkod. 30461133 Leverantör LoGiCal Leverantörsnummer LGCFOR0083.04
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl (2-Oxopyrrolidin-1-yl)acetate
CAS 59776-88-4
Analyt- eller komponentnamn Methyl (2-Oxopyrrolidin-1-yl)acetate
Frakt skick Room Temperature
Molekylformel C7 H11 N O3
Certifikat och efterlevnad ISO 9001
Synonym Methyl (2-Oxopyrrolidin-1-yl)acetate, Piracetam Imp. B (EP), 2-Oxo-1-pyrrolidineacetic acid methyl ester, (2-Oxopyrrolidin-1-yl)acetic acid methyl ester, 1-(Methoxycarbonylmethyl)-2-pyrrolidone, 2-Ketopyrrolidine-1-acetic acid methyl ester, Methyl 2-(2-oxo-1-pyrrolidinyl)acetate, Methyl 2-oxo-1-pyrrolidineacetate, Methyl 2-pyrrolidinon-1-ylacetate, Methyl 2-pyrrolidone-1-acetate, Piracetam Impurity B
InChI formel InChI=1S/C7H11NO3/c1-11-7(10)5-8-4-2-3-6(8)9/h2-5H2,1H3
Lösningstyp Neat
LEDER COC(=O)CN1CCCC1=O
IUPAC-namn methyl 2-(2-oxopyrrolidin-1-yl)acetate
Industry Type Clinical Testing
Molekylvikt (g/mol) 157.17
Kvantitet 10 mg
Formel vikt 157.0739 g/mol