Läs mer
Methyl 2-(p-Tolyl)benzoate, TRC
Produktkod. 30465037
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30465037 5 g
1 options
Produktkod. 30465037 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3318605G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 2-(p-Tolyl)benzoate
CAS 114772-34-8
Molekylformel C15H14O2
Kvantitet 5 g
Synonym 4'-Methyl-[1,1'-biphenyl]-2-carboxylic Acid Methyl Ester, Methyl 2-(4-Methylphenyl)benzoate, Methyl 4-Methyl-2'-biphenylcarboxylate, Methyl 4'-Methylbiphenyl-2-carboxylate
InChI formel InChI=1S/C15H14O2/c1-11-7-9-12(10-8-11)13-5-3-4-6-14(13)15(16)17-2/h3-10H,1-2H3
LEDER COC(=O)c1ccccc1c2ccc(C)cc2
IUPAC-namn methyl 2-(4-methylphenyl)benzoate
Molekylvikt (g/mol) 226.27
Formel vikt 226.0994