Läs mer
Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate, TRC
Produktkod. 30474684
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474684 50 mg
30473720 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30474684 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33001050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57113-90-3
Molekylformel C13H16N2O6
Molekylvikt (g/mol) 296.28
Synonym 2-[[(1,1-Dimethylethoxy)carbonyl]amino]-3-nitrobenzoic Acid Methyl Ester
IUPAC-namn methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-nitrobenzoate
LEDER COC(=O)c1cccc(c1NC(=O)OC(C)(C)C)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material Methyl 2-(tert-Butoxycarbonylamino)-3-nitrobenzoate
Kvantitet 50 mg
InChI formel InChI=1S/C13H16N2O6/c1-13(2,3)21-12(17)14-10-8(11(16)20-4)6-5-7-9(10)15(18)19/h5-7H,1-4H3,(H,14,17)
Formel vikt 296.1008