missing translation for 'onlineSavingsMsg'
Learn More

Methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropionate, 98%, Thermo Scientific Chemicals

Catalog Number 15466666
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
32981-85-4
C17H17NO4
299.326
MFCD00673331
UYJLJICUXJPKTB-LSDHHAIUSA-N
2r,3s-methyl 3-benzamido-2-hydroxy-3-phenylpropanoate, methyl 2r,3s-3-benzoylamino-2-hydroxy-3-phenylpropanoate, bz-rs-iser 3-ph-ome, benzenepropanoic acid, beta-benzoylamino-alpha-hydroxy-, methyl ester, alphar,betas, methyl 2r,3s-3-benzamido-2-hydroxy-3-phenylpropanoate, 2r,3s-n-tert-butoxycarbony-3-phenylisoserine methyl ester, methyl 2r,3s-2-hydroxy-3-phenyl-3-phenylformamido propanoate, pubchem9699, n-benzoyl-2r,3s-3-phenylisoserine methyl ester, methyl 2r,3s-n-benzoylphenylisoserinate
182104
methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropanoate
COC(=O)C(C(C1=CC=CC=C1)NC(=O)C2=CC=CC=C2)O
This item is not returnable. View return policy
32981-85-4
C17H17NO4
299.326
MFCD00673331
UYJLJICUXJPKTB-LSDHHAIUSA-N
2r,3s-methyl 3-benzamido-2-hydroxy-3-phenylpropanoate, methyl 2r,3s-3-benzoylamino-2-hydroxy-3-phenylpropanoate, bz-rs-iser 3-ph-ome, benzenepropanoic acid, beta-benzoylamino-alpha-hydroxy-, methyl ester, alphar,betas, methyl 2r,3s-3-benzamido-2-hydroxy-3-phenylpropanoate, 2r,3s-n-tert-butoxycarbony-3-phenylisoserine methyl ester, methyl 2r,3s-2-hydroxy-3-phenyl-3-phenylformamido propanoate, pubchem9699, n-benzoyl-2r,3s-3-phenylisoserine methyl ester, methyl 2r,3s-n-benzoylphenylisoserinate
182104
methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropanoate
COC(=O)C(C(C1=CC=CC=C1)NC(=O)C2=CC=CC=C2)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 178°C to 183°C
Color White
Quantity 250 mg
Formula Weight 299.32
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropionate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'