Läs mer
Methyl (2RS)-(2-Chlorophenyl)[4,7-dihydrothieno[2,3-c]pyridin-6(5H)-yl]acetate Hydrogen Sulfate, Mikromol™
Beskrivning
Methyl (2RS)-(2-Chlorophenyl)[4,7-dihydrothieno[2,3-c]pyridin-6(5H)-yl]acetate Hydrogen Sulfate, Mikromol
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl (2RS)-(2-Chlorophenyl)[4,7-dihydrothieno[2,3-c]pyridin-6(5H)-yl]acetate Hydrogen Sulfate |
| CAS | 2137493-39-9 |
| Analyt- eller komponentnamn | Methyl (2RS)-(2-Chlorophenyl)[4,7-dihydrothieno[2,3-c]pyridin-6(5H)-yl]acetate Hydrogen Sulfate |
| Frakt skick | Room Temperature |
| Molekylformel | C16 H16 Cl N O2 S . H2 O4 S |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Clopidogrel Hydrogen Sulfate Imp. B (EP), Clopidogrel Imp. B (EP), Clopidogrel USP Related Compound B, Clopidogrel USP RC B, Methyl (2RS)-(2-Chlorophenyl)[4,7-dihydrothieno[2,3-c]pyridin-6(5H)-yl]acetate Hydrogen Sulfate, Clopidogrel Hydrogen Sulfate Imp. B (EP) as Racemate Hydrogen Sulfate, Clopidogrel Related Compound B as Sulfate, Clopidogrel Hydrogen Sulfate Impurity B as racemic Hydrogen Sulfate, Clopidogrel Impurity B as racemic Hydrogen Sulfate |
| InChI formel | InChI=1S/C16H16ClNO2S.H2O4S/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-11-7-9-21-14(11)10-18;1-5(2,3)4/h2-5,7,9,15H,6,8,10H2,1H3;(H2,1,2,3,4) |
| Lösningstyp | Neat |
| LEDER | COC(=O)C(N1CCc2ccsc2C1)c3ccccc3Cl.OS(=O)(=O)O |
| Visa mer |