Läs mer
Methyl (2S)-2-amino-2-(4-hydroxyphenyl)acetate Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methyl (2S)-Amino(4-hydroxyphenyl)ethanoate Hydrochloride |
| CAS | 127369-30-6 |
| Molekylformel | C9 H11 N O3 . Cl H |
| Kvantitet | 250 mg |
| Synonym | Benzeneacetic acid, α-amino-4-hydroxy-, methyl ester, hydrochloride (1:1), (αS)-, Benzeneacetic acid, α-amino-4-hydroxy-, methyl ester, hydrochloride, (S)-, Benzeneacetic acid, α-amino-4-hydroxy-, methyl ester, hydrochloride, (αS)- (9CI), (S)-Methyl 2-amino-2-(4-hydroxyphenyl)acetate hydrochloride, Methyl (2S)-amino(4-hydroxyphenyl)ethanoate hydrochloride |
| InChI formel | InChI=1S/C9H11NO3.ClH/c1-13-9(12)8(10)6-2-4-7(11)5-3-6;/h2-5,8,11H,10H2,1H3;1H/t8-;/m0./s1 |
| LEDER | Cl.COC(=O)[C@@H](N)c1ccc(O)cc1 |
| IUPAC-namn | methyl (2S)-2-amino-2-(4-hydroxyphenyl)acetate;hydrochloride |
| Molekylvikt (g/mol) | 217.6495 |
| Formel vikt | 217.0506 |