Läs mer
Methyl (2S)-2-Aminobutanoate, TRC
Produktkod. 30478745
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30478745 1 g
1 options
Produktkod. 30478745 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2857351G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl (2S)-2-Aminobutanoate
CAS 15399-22-1
Molekylformel C5 H11 N O2
Kvantitet 1 g
Synonym Butanoic acid','2-amino-','methyl ester','(2S)-, Butanoic acid','2-amino-','methyl ester','(S)-, Butyric acid','2-amino-','methyl ester','L- (8CI), Methyl (2S)-2-aminobutanoate, (S)-2-Aminobutanoic acid methyl ester, (S)-Methyl 2-aminobutanoate, L-2-Aminobutyric a
InChI formel InChI=1S/C5H11NO2/c1-3-4(6)5(7)8-2/h4H,3,6H2,1-2H3/t4-/m0/s1
LEDER CC[C@H](N)C(=O)OC
IUPAC-namn methyl (2S)-2-aminobutanoate
Molekylvikt (g/mol) 117.146
Formel vikt 117.079