Läs mer
Methyl 3-(2-Aminoethyl)benzoate Hydrochloride, TRC
Produktkod. 30318093
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30318093 100 mg
30317686 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30318093 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA608845100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3-(2-Aminoethyl)benzoate Hydrochloride
CAS 167846-36-8
Molekylformel C10H13NO2 . HCl
Kvantitet 100 mg
Synonym 3-(2-Aminoethyl)-benzoic Acid Methyl Ester Hydrochloride
InChI formel InChI=1S/C10H13NO2.ClH/c1-13-10(12)9-4-2-3-8(7-9)5-6-11;/h2-4,7H,5-6,11H2,1H3;1H
LEDER COC(=O)c1cccc(CCN)c1
IUPAC-namn methyl 3-(2-aminoethyl)benzoate;hydrochloride
Molekylvikt (g/mol) 215.67