Läs mer
Methyl 3-[[[2-[(Diaminomethylene]amino-4-thiazolyl]methyl]-thio]propionimidate Hyxdrochloride, TRC
Produktkod. 30478852
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30478852 250 mg
1 options
Produktkod. 30478852 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM299525250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3-((2-((Diaminomethylene)amino-4-thiazolyl)methyl)-thio)propionimidate Hyxdrochloride
CAS 1797843-63-0
Molekylformel C9H15N5OS2 . HCl
Kvantitet 250 mg
Synonym 3-[[[2-[(Aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]propanimidic Acid Methyl Ester Hydrochloride
InChI formel InChI=1S/C9H15N5OS2.ClH/c1-15-7(10)2-3-16-4-6-5-17-9(13-6)14-8(11)12;/h5,10H,2-4H2,1H3,(H4,11,12,13,14);1H
LEDER Cl.COC(=N)CCSCc1csc(N=C(N)N)n1
IUPAC-namn methyl 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]propanimidate;hydrochloride
Molekylvikt (g/mol) 309.8
Formel vikt 309.0485