Läs mer
Methyl 3-[2-(Isopropylamino)propanamido]-4-methylthiophene-2-carboxylate, TRC
Produktkod. 30134261
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134261 50 mg
30135071 100 mg
30009924 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134261 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM31451550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1796888-45-3
Molekylformel C13 H20 N2 O3 S
Molekylvikt (g/mol) 284.37
Synonym 2-Thiophenecarboxylic acid, 4-methyl-3-[[2-[(1-methylethyl)amino]-1-oxopropyl]amino]-, methyl ester, Methyl 4-Methyl-3-[[(2RS)-2-[(1-methylethyl)amino]propanoyl]amino]thiophene-2-carboxylate, Articaine Hydrochloride Imp. E (EP), Isopropylarticaine
IUPAC-namn methyl 4-methyl-3-[2-(propan-2-ylamino)propanoylamino]thiophene-2-carboxylate
LEDER COC(=O)c1scc(C)c1NC(=O)C(C)NC(C)C
Specifikationer
Kemiskt namn eller material Isopropylarticaine
Kvantitet 50 mg
InChI formel InChI=1S/C13H20N2O3S/c1-7(2)14-9(4)12(16)15-10-8(3)6-19-11(10)13(17)18-5/h6-7,9,14H,1-5H3,(H,15,16)
Formel vikt 284.1195