missing translation for 'onlineSavingsMsg'
Läs mer

Methyl 3-((2R)-2-Hydroxy-4-(((((S)-1-methoxy-1-oxopropan-2-yl)amino)(phenoxy)phosphoryl)oxy)-3,3-dimethylbutanamido)propanoate (Mixture of Diastereomers), TRC

Produktkod. 30478742
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30478742 25 mg
30467021 10 mg
30467854 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30478742 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM31216025MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1858268-66-2
Molekylformel C20H31N2O9P
Molekylvikt (g/mol) 474.44
Synonym Fosmetpantotenate, RE 024, Dimethyl 8-hydroxy-2,7,7-trimethyl-4,9-dioxo-4-phenoxy-5-oxa-3,10-diaza-4λ5-phosphatridecanedioate,mixture of (2S,4R,8R) and (2S,4S,8R) Epimers
LEDER COC(=O)CCNC(=O)[C@H](O)C(C)(C)COP(=O)(N[C@@H](C)C(=O)OC)Oc1ccccc1

Specifications

Kemiskt namn eller material Methyl 3-((2R)-2-Hydroxy-4-(((((S)-1-methoxy-1-oxopropan-2-yl)amino)(phenoxy)phosphoryl)oxy)-3,3-dimethylbutanamido)propanoate (Mixture of Diastereomers)
Kvantitet 25 mg
InChI formel InChI=1S/C20H31N2O9P/c1-14(19(26)29-5)22-32(27,31-15-9-7-6-8-10-15)30-13-20(2,3)17(24)18(25)21-12-11-16(23)28-4/h6-10,14,17,24H,11-13H2,1-5H3,(H,21,25)(H,22,27)/t14-,17-,32?/m0/s1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.