Läs mer
Methyl 3-(3’,4’-Dimethoxyphenyl)propenoate, TRC
Produktkod. 30483177
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30483177 5 g
1 options
Produktkod. 30483177 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3028705G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3-(3',4'-Dimethoxyphenyl)propenoate
CAS 5396-64-5
Molekylformel C12 H14 O4
Kvantitet 5 g
Synonym Methyl 3,4-Dimethoxycinnamate, 3-(3,4-Dimethoxyphenyl)-2-propenoic Acid Methyl Ester, 3,4-O-Dimethylcaffeic Acid Methyl Ester, NSC 4331
InChI formel InChI=1S/C12H14O4/c1-14-10-6-4-9(8-11(10)15-2)5-7-12(13)16-3/h4-8H,1-3H3
LEDER COC(=O)C=Cc1ccc(OC)c(OC)c1
IUPAC-namn methyl 3-(3,4-dimethoxyphenyl)prop-2-enoate
Molekylvikt (g/mol) 222.24
Formel vikt 222.0892