Läs mer
Methyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate, TRC
Produktkod. 30476820
Change view
Kvantitet:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476820 5 g
30465599 25 g
30469121 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30476820 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3011785G
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6386-38-5
Molekylformel C18 H28 O3
Molekylvikt (g/mol) 292.41
Synonym Hydrocinnamic acid, 3,5-di-tert-butyl-4-hydroxy-, methyl ester (7CI,8CI), 3,5-Bis(1,1-dimethylethyl)-4-hydroxybenzenepropanoic acid methyl ester, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid methyl ester, Fenozan 1, Methyl (3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Methyl 3,5-di-tert-butyl-4-hydroxy-β-phenylpropionate, Methyl 3,5-di-tert-butyl-4-hydroxyhydrocinnamate, Methyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate, Methyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Methyl 3-(3',5'-di-tert-butyl-4'-hydroxyphenyl)propionate, Methyl 3-(4-hydroxy-3,5-di-tert-butylphenyl)propionate, Methyl β-(4-hydroxy-3,5-di-tert-butylphenyl)propanoate, Methyl β-(4-hydroxy-3,5-di-tert-butylphenyl)propionate, Methyl β-3,5-di-tert-butyl-4-hydroxyphenylpropionate, Methylox, Metilox, Phenosan methyl ester, Ralox 35, Benzenepropanoic acid, 3,5-bis(1,1-dimethylethyl)-4-hydroxy-, methyl ester
IUPAC-namn methyl 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate
LEDER COC(=O)CCc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C
Specifikationer
Kemiskt namn eller material Metilox
Kvantitet 5 g
InChI formel InChI=1S/C18H28O3/c1-17(2,3)13-10-12(8-9-15(19)21-7)11-14(16(13)20)18(4,5)6/h10-11,20H,8-9H2,1-7H3
Formel vikt 292.2038