Läs mer
Methyl 3,4,5-Trimethoxycinnamate (>85%), TRC
Produktkod. 30484040
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30484040 100 mg
1 options
Produktkod. 30484040 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM219425100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3,4,5-Trimethoxycinnamate
CAS 20329-96-8
Molekylformel C13 H16 O5
Kvantitet 100 mg
Synonym 2-Propenoic acid, 3-(3,4,5-trimethoxyphenyl)-, methyl ester, (2E)- (9CI, ACI), 2-Propenoic acid, 3-(3,4,5-trimethoxyphenyl)-, methyl ester, (E)- (ZCI), Cinnamic acid, 3,4,5-trimethoxy-, methyl ester, (E)- (8CI), Methyl (E)-3,4,5-trimethoxycinnamate, Methyl 3,4,5-trimethoxy-trans-cinnamate, Methyl trans-3,4,5-trimethoxycinnamate, Methyl trans-3-(3,4,5-trimethoxyphenyl)acrylate, trans-3-(3,4,5-Tri-methoxyphenyl-3-Propenoic acid methyl ester
InChI formel InChI=1S/C13H16O5/c1-15-10-7-9(5-6-12(14)17-3)8-11(16-2)13(10)18-4/h5-8H,1-4H3/b6-5+
LEDER COC(=O)\C=C\c1cc(OC)c(OC)c(OC)c1
IUPAC-namn methyl (E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoate
Molekylvikt (g/mol) 252.26
Formel vikt 252.0998