Läs mer
Methyl 3,4,5-Tris(benzyloxy)benzoate, TRC
Produktkod. 30476427
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30476427 2.5 g
1 options
Produktkod. 30476427 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3312852.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3,4,5-Tris(benzyloxy)benzoate
CAS 70424-94-1
Molekylformel C29H26O5
Kvantitet 2.5 g
Synonym 3,4,5-Tris(phenylmethoxy)benzoic Acid Methyl Ester, Methyl 3,4,5-Tribenzyloxybenzoate, Methyl Tri-O-benzylgallate, NSC 525276
InChI formel InChI=1S/C29H26O5/c1-31-29(30)25-17-26(32-19-22-11-5-2-6-12-22)28(34-21-24-15-9-4-10-16-24)27(18-25)33-20-23-13-7-3-8-14-23/h2-18H,19-21H2,1H3
LEDER COC(=O)c1cc(OCc2ccccc2)c(OCc3ccccc3)c(OCc4ccccc4)c1
IUPAC-namn methyl 3,4,5-tris(phenylmethoxy)benzoate
Molekylvikt (g/mol) 454.51
Formel vikt 454.178