Läs mer
Methyl 3,4-Di-O-acetyl-Beta-L-idopyranosiduronate 1,2-(Methylorthoacetate), TRC
Produktkod. 30483333
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30483333 10 mg
1 options
Produktkod. 30483333 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM29867510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3,4-Di-O-acetyl-beta-L-idopyranosiduronate 1,2-(Methylorthoacetate)
CAS 107952-66-9
Molekylformel C14H20O10
Kvantitet 10 mg
Synonym β-L-Idopyranuronic acid','1,2-O-(1-methoxyethylidene)-','methyl ester','diacetate (9CI), 5H-1,3-Dioxolo[4,5-b]pyran','β-L-idopyranuronic acid deriv., Methyl 3,4-di-O-acetyl-β-L-idopyranosiduronate 1,2-(methylorthoacetate), 3,4-Di-O-acetyl-1,2-O-(1-methoxyethylidene)-β-L-idopyranuronic acid methyl ester
InChI formel InChI=1S/C14H20O10/c1-6(15)20-8-9(21-7(2)16)11-13(22-10(8)12(17)18-4)24-14(3,19-5)23-11/h8-11,13H,1-5H3/t8-,9-,10+,11+,13+,14?/m0/s1
LEDER COC(=O)[C@@H]1O[C@@H]2OC(C)(OC)O[C@@H]2[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC-namn methyl (3aR,5R,6S,7S,7aR)-6,7-diacetyloxy-2-methoxy-2-methyl-5,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-b]pyran-5-carboxylate
Molekylvikt (g/mol) 348.3
Formel vikt 348.1056