Läs mer
Methyl 3,5-Dinitrosalicylate, TRC
Produktkod. 30467504
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30467504 250 mg
1 options
Produktkod. 30467504 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM303775250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3,5-Dinitrosalicylate
CAS 22633-33-6
Molekylformel C8H6N2O7
Kvantitet 250 mg
Synonym 3,5-Dinitro Salicylic Acid Methyl Ester, NSC 203306
InChI formel InChI=1S/C8H6N2O7/c1-17-8(12)5-2-4(9(13)14)3-6(7(5)11)10(15)16/h2-3,11H,1H3
LEDER COC(=O)c1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-]
IUPAC-namn methyl 2-hydroxy-3,5-dinitrobenzoate
Molekylvikt (g/mol) 242.17
Formel vikt 242.0175