Läs mer
Methyl 3'-Amino-1,1'-biphenyl-3-carboxylate, TRC
Produktkod. 30476802
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30476802 500 mg
1 options
Produktkod. 30476802 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM293265500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3'-Amino-1,1'-biphenyl-3-carboxylate
CAS 168619-25-8
Molekylformel C14 H13 N O2
Kvantitet 500 mg
Synonym 3'-Amino-[1,1'-biphenyl]-3-carboxylic Acid Methyl Ester, Methyl 3'-Amino-1,1'-biphenyl-3-carboxylate, Methyl 3'-Aminobiphenyl-3-carboxylate
InChI formel InChI=1S/C14H13NO2/c1-17-14(16)12-6-2-4-10(8-12)11-5-3-7-13(15)9-11/h2-9H,15H2,1H3
LEDER COC(=O)c1cccc(c1)c2cccc(N)c2
IUPAC-namn methyl 3-(3-aminophenyl)benzoate
Molekylvikt (g/mol) 227.258
Formel vikt 227.095