Läs mer
Methyl 3β-Amino-7α,12α-dihydroxy-5β-cholan-24-oate, TRC
Produktkod. 30483119
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30483119 100 mg
1 options
Produktkod. 30483119 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM286025100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 3Beta-Amino-7Alpha,12Alpha-dihydroxy-5Beta-cholan-24-oate
CAS 135053-62-2
Molekylformel C25 H43 N O4
Kvantitet 100 mg
Synonym Methyl 3β-Aminocholic Acid Ester, Methyl 3β-Aminocholate, Methyl (3β,5β,7α,12α)-3-Amino-7,12-dihydroxy-cholan-24-oic Acid Ester
InChI formel InChI=1S/C25H43NO4/c1-14(5-8-22(29)30-4)17-6-7-18-23-19(13-21(28)25(17,18)3)24(2)10-9-16(26)11-15(24)12-20(23)27/h14-21,23,27-28H,5-13,26H2,1-4H3/t14-,15+,16+,17-,18+,19+,20-,21+,23+,24+,25-/m1/s1
LEDER COC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@@H](N)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
IUPAC-namn methyl (4R)-4-[(3S,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3-amino-7,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
Molekylvikt (g/mol) 421.613
Formel vikt 421.319